Learn More
Biomatik Corporation Recombinant Human Desmoglein-3 (DSG3), partial
Supplier:  Biomatik Corporation RPC25732
Encompass_Preferred
Recombinant Human Desmoglein-3 (DSG3), partial is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: DSG3. Target Synonyms: 130 kDa pemphigus vulgaris antigen (PVA; Cadherin family member 6; CDHF6). Accession Number: P32926. Expression Region: 50~615aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 67.0kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5864
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.