Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human DLL4 Fc Chimera Biotinylated Protein, CF
SDP

Catalog No. BT10185050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BT10185050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BT10185050 Supplier R&D Systems™ Supplier No. BT10185050
Add to Cart
Add to Cart

Recombinant Protein

Measured by its binding ability in a functional ELISA. Biotinylated Recombinant Human DLL4 Fc Chimera (Catalog # BT10185) binds to Recombinant Human Notch-1 Fc Chimera (Catalog # 3647-TK ) with an ED50 of 7.00-84.0 ng/mL.

Specifications

For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 54567
Molecular Weight (g/mol) Observed: 83-98 kDa, under reducing conditions.,Theoretical: 81 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human DLL4 protein Human DLL4 (Ser27-Pro524) (N-terminus) Accession # Q9NR61 IEGRMDP Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias delta 4, Delta 4 precursor, delta ligand 4, delta4, delta-like 4 (Drosophila), delta-like 4 homolog, delta-like 4 homolog (Drosophila), delta-like 4 protein, delta-like protein 4, Drosophila Delta homolog 4, hdelta2, MGC126344, notch ligand delta-2, notch ligand DLL4
Conjugate Biotin
Reconstitution Reconstitute at 250 μg/mL in water.
Protein DLL4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.