Learn More
Description
Specifications
Specifications
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 1803 |
| Molecular Weight (g/mol) | M.W.-observed: 116-131 kDa, under reducing conditions, M.W.-theroretical: 112 kDa |
| Quantity | 20 μg |
| Source | Human embryonic kidney cell, HEK293-derived human DPPIV/CD26 protein Human CD26 (Asn29-Pro766) (C-terminus) Accession # CAA43118.1 DIEGRMD Human IgG1 (Pro100-Lys330) (N-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2 TP103, CD26, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPP4, DPPIV, EC 3.4.14.5 |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
