Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Dystroglycan Fc Chimera Protein
SDP

Catalog No. 10223DG050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-223-DG050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-223-DG050 Supplier R&D Systems™ Supplier No. 10223DG050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measured by the ability of the immobilized protein to enhance the adhesion of H4 human neuroglioma cells. The ED50 for this effect is 1.5 to 9 μg/mL.

Specifications

Accession Number Q14118
For Use With (Application) Bioactivity
Gene ID (Entrez) 1605
Molecular Weight (g/mol) 68 kDa (alpha subunit) and 37 kDa (beta subunit)
Name A3a, AGRNR, Dag-1, DAG1, DAG156DAG, dystroglycan, dystroglycan 1 (dystrophin-associated glycoprotein 1), Dystrophin-associated glycoprotein 1
Quantity 50 μg
Source

Human embryonic kidney cell, HEK293-derived human Dystroglycan protein

Human Dystroglycan (Gln28-Val749) Accession # Q14118IEGRMDHuman IgG1 (Pro100-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 2 weeks, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 500 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.