Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human E-Selectin/CD62E His-tag Protein

Catalog No. 10258ES100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-258-ES100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-258-ES100 Supplier R&D Systems™ Supplier No. 10258ES100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measured by the ability of the immobilized protein to support the adhesion of U937 human histiocytic lymphoma cells.

ED50 for this effect is 0.2 to 1 μg/mL

Specifications

Accession Number P16581
For Use With (Application) Bioactivity
Gene ID (Entrez) 6401
Molecular Weight (g/mol) 84 to 109 kDa, under reducing conditions
Quantity 100 μg
Source Chinese Hamster Ovary cell line, CHO-derived human E-Selectin/CD62E protein Trp22-Pro556, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration < 0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 500 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS
Purity or Quality Grade > 95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.