Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human EBI3 Protein
SDP

Catalog No. 11084EI050 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11084EI050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11084EI050 Supplier R&D Systems™ Supplier No. 11084EI050

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Measured by its binding ability in a functional ELISA. When Recombinant Human EBI3 Protein (Catalog # 11084-EI) is immobilized at 5.0 μg/mL (100 μL/well), the concentration of Recombinant Mouse IL-27 p28/IL-30 Protein (Catalog # 7430-ML ) that produces 50% of the optimal binding response is 40.0-300 ng/mL.

Specifications

Accession Number Q14213.2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 10148
Molecular Weight (g/mol) MolecularWeight-observed: 30-40 kDa, under reducing conditions., MolecularWeight-theroretical: 23 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human EBI3 protein Arg21-Lys220
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias cytokine receptor, EBI-3, EBV-induced gene 3 protein, Epstein-Barr virus induced 3, Epstein-Barr virus induced gene 3, Epstein-Barr virus-induced gene 3 protein, IL-27 beta, IL-27 subunit beta, IL27B, IL-27 B, IL35 EBI3, IL-35 EBI3, interleukin-27 subunit beta
Conjugate Unconjugated
Reconstitution Reconstitute at 200 μg/mL in PBS.
Product Line R&D Systems
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein EBI3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.