Learn More
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Accession Number | P01133 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 1950 |
| Molecular Weight (g/mol) | M.W. (Observed): 6 kDa, reducing conditions; M.W. (theoretical): 6 kDa |
| Name | EGF |
| Quantity | 1 mg |
| Source | E. coli-derived human EGF protein Asn971-Arg1023, with an N-terminal Met |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

