Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human EGFR Fc Chimera Alexa Fluor™ 488 Protein

Catalog No. AFG344020
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AFG344020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AFG344020 Supplier R&D Systems™ Supplier No. AFG344020
Add to Cart
Add to Cart
This item is not returnable. View return policy
Measured by flow cytometry for its ability to bind anti-human EGFR Monoclonal Antibody conjugated beads. The concentration of Recombinant Human EGFR Fc-tag Alexa Fluor™ 488 (Catalog # AFG344) that produces 50% of the binding response is 8.00-120 ng/mL.

Specifications

Accession Number CAA25240.1
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 1956
Molecular Weight (g/mol) M.W.-observed: 118-134 kDa, under reducing conditions., M.W.-theroretical: 95 kDa (monomer)
Quantity 20 μg
Source Mouse myeloma cell line, NS0-derived human EGFR protein Human EGFR (Leu25-Ser645) (C-terminus) Accession # CAA25240.1 IEGRMD Human IgG1 (Pro100-Lys330) (N-terminus)
Storage Requirements Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, EC 2.7.10, EC 2.7.10.1, EGF R, epidermal growth factor receptor, epidermal growth factor receptor (avian erythroblastic leukemia viral (v-erb-b)oncogene homolog), ErbB, ErbB1, ERBB1PIG61, HER1, HER-1, mENA, Proto-oncogene c-ErbB-1, Receptor tyrosine-protein kinase erbB-1
Conjugate Alexa Fluor 488
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein EGFR
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.