Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human eIF5A Protein
SDP

Catalog No. NBC121053 Shop All R&D Systems Products
Change view
Click to view available options
0.1mg; Unlabeled
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NBC121053 0.1mg; Unlabeled
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC121053 Supplier Novus Biologicals™ Supplier No. NBC121053
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-154 of Human eIF5A The Recombinant Human eIF5A Protein is derived from E. coli. The Recombinant Human eIF5A Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_001961
For Use With (Application) ELISA, SDS-PAGE
Formulation 50mM Tris-HCl buffer (pH 7.5) containing 10% glycerol
Gene ID (Entrez) 1984
Molecular Weight (g/mol) 16.8kDa
Name eIF5A Protein
Purification Method Affinity purified
Immunogen MADDLDFETG DAGASATFPM QCSALRKNGF VVLKGRPCKI VEMSTSKTGK HGHAKVHLVG IDIFTGKKYE DICPSTHNMD VPNIKRNDFQ LIGIQDGYLS LLQDSGEVRE DLRLPEGDLG KEIEQKYDCG EEILITVLSA MTEEAAVAIK AMAK
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.