Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human EMMPRIN/CD147 Fc Chimera (NS0) Protein

Catalog No. 972EMN050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
972EMN050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 972EMN050 Supplier R&D Systems™ Supplier No. 972EMN050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Isoform 2. aa 22-205

Measured by the ability of the immobilized protein to induce active MMP-1 secretion by NHLF human normal lung fibroblasts. The ED50 for this effect is 2-8 μg/mL. Measured by its binding ability in a functional ELISA with Recombinant SARS-CoV-2 Spike RBD Fc Chimera (Catalog # 10499-CV ).

Specifications

Accession Number NP_001309172.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 682
Molecular Weight (g/mol) M.W. (Observed): 60 - 65 kDa, under reducing conditions.
Name EMMPRIN/CD147 Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human EMMPRIN/CD147 protein Human EMMPRIN/CD147 (Thr25-His205) (C-terminus) Accession # NP_001309172.1 DIEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (N-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol BSG
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein EMMPRIN/CD147
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.