Learn More
Description
Specifications
Specifications
| Gene ID (Entrez) | 2021 |
| Species | Human |
| Product Line | Novus |
| Purity | >80% by SDS-PAGE and Coomassie blue staining |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Gene Alias | EC 3.1.30.-, Endo G, endonuclease G, endonuclease G, mitochondrial, FLJ27463, mitochondrial endonuclease G |
| Gene Symbol | ENDOG |
| Molecular Weight (g/mol) | 53.02 kDa |
| Quantity | 2 μg |
| Research Category | Apoptosis, Cancer, Hypoxia |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
