Learn More
Description
Order Info
The product is shipped at ambient temperature. Upon receipt, store it immediately at the recommended storage temperatures.
Specifications
Specifications
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived human Endoglycan/PODXL2 protein Gly33-Thr500,with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
