Learn More
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human VEGF R1/Flt-1 (Catalog # 321-FL ) is coated at 5μg/mL (100μL/well), the concentration of Recombinant Human Endorepellin/Perlecan that produces 50% of the optimal binding response is approximately 0.75-6μg/mL.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 3339 |
| Molecular Weight (g/mol) | 76.7 kDa |
| Name | Endorepellin/Perlecan |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived human Endorepellin/Perlecan protein Arg3684-Ser4391 (Ser4004Thr),with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | basement membrane-specific heparan sulfate proteoglycan core protein, endorepellin (domain V region), heparan sulfate proteoglycan 2, HSPG, perlecan, perlecan proteoglycan, PLCSchwartz-Jampel syndrome 1 (chondrodystrophic myotonia), PRCAN, SJA, SJS, SJS1 |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
