Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Enolase 1 Protein
SDP

Catalog No. NBP130262 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP130262 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP130262 Supplier Novus Biologicals™ Supplier No. NBP130262
Add to Cart
Add to Cart

Highly purified and high bioactivity. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-434 of Human Enolase 1 The Recombinant Human Enolase 1 Protein is derived from E. coli. The Recombinant Human Enolase 1 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Accession Number P06733
For Use With (Application) Bioactivity, Microarray, SDS-PAGE, Western Blot
Formulation 20mM Tris-HCl buffer (pH 7.5), 1mM MgSO4, 10% glycerol
Gene ID (Entrez) 2023
Molecular Weight (g/mol) 47kDa
Name Enolase 1 Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen Alpha-enolase, 1-434aa, Human, E.coli (NP_001419).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.