Learn More
R&D Systems™ Recombinant Human Enolase 2 His-tag Protein
Description
Specifications
Specifications
| Accession Number | P09104.3 |
| Formulation | Supplied as a 0.2 μm filtered solution in MES, NaCl, KCl and MgSO4. |
| Gene ID (Entrez) | 2026 |
| Molecular Weight (g/mol) | Observed: 46 kDa, under reducing conditions |
| Quantity | 50 μg |
| Source | E. coli-derived human Enolase 2/Neuron-specific Enolase protein Met1-Leu434 with an N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Purity or Quality Grade | >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.