Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human ENPP-5 Protein
SDP

Catalog No. 8655EN020 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8655EN020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8655EN020 Supplier R&D Systems™ Supplier No. 8655EN020

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to cleave O-(4-Nitrophenylphosphoryl) choline. The specific activity is >300 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number Q9UJA9
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 59084
Molecular Weight (g/mol) M.W. (Observed): 70-80 kDa, reducing conditions; M.W. (theoretical): 48 kDa
Quantity 20 μg
Source Human embryonic kidney cell, HEK293-derived human ENPP-5 protein Pro25-Ser430, with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias EC 3.1, ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative function), ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative), ectonucleotide pyrophosphatase/phosphodiesterase family member 5, E-NPP 5, ENPP5, KIAA0879, NPP5, NPP-5
Common Name ENPP-5
Gene Symbol ENPP5
Biological Activity Measured by its ability to cleave O-(4-Nitrophenylphosphoryl) choline. The specific activity is >300 pmol/min/μg, as measured under the described conditions.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Pro25-Ser430, with a C-terminal 10-His tag
Product Line R&D Systems
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane)
Protein ENPP-5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.