Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Recombinant Human ENPP1/PC1 Protein, >95% Purity, 100 μg

Catalog No. 502582635
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-2635 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-2635 Supplier Abcam Supplier No. AB167943100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human ENPP1/PC1 protein expressed in HEK293 cells with >95% purity.

  • Protein Name: ENPP1/PC1
  • Source: Human, expressed in HEK 293 cells
  • Sequence: 98 to 925 aa fragment
  • Purity: >95%
  • Endotoxin Level: < 1 EU/μg
  • Applications: ELISA, SDS-PAGE, WB
  • Form: Lyophilized; Constituents: 99% Phosphate Buffer, 0.43% Sodium chloride

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule ENPP1/PC1 (Human)
Content And Storage Store at -80°C.
Form Lyophilized
Buffer 99% phosphate buffer, 0.43% sodium chloride
Shipping Condition Blue Ice
Cross Reactivity Human
Formulation Lyophilized from 99% Phosphate Buffer, 0.43% Sodium chloride
Quantity 100 μg
For Use With (Application) SDS-PAGE, ELISA, Western Blot
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.