Learn More
R&D Systems™ Recombinant Human Enteropeptidase/Enterokinase Protein
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Accession Number | P98073 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl and CaCl2. |
| Gene ID (Entrez) | 5651 |
| Molecular Weight (g/mol) | M.W. (Observed): 98-117 kDa (heavy chain) & 46-49 kDa (light chain), reducing conditions; M.W. (theoretical): 67, 55, 53 kDa (heavy chain) & 27 kDa (light chain) |
| Quantity | 20 μg |
| Source | Mouse myeloma cell line, NS0-derived human Enteropeptidase/Enterokinase protein Leu41-His1019 with a C-terminal 9-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.