Learn More
R&D Systems™ Recombinant Human EphA1 Fc Chimera Protein
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in MES, NaCl, PEG, CHAPS and Imidazole. |
| Gene ID (Entrez) | 2041 |
| Molecular Weight (g/mol) | 83.5 kDa (monomer) |
| Name | EphA1 Fc Chimera |
| Quantity | 100 μg |
| Source | Human embryonic kidney cell,HEK293-derived human EphA1 protein, (N-terminus) Human EphA1 (Met1-Glu547) Accession &Num; AAA36747 DIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. 3 months, -20 to -70° C under sterile conditions after opening. |
| Structural Form | Disulfide-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.