Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human EphB6 Fc Chimera Protein

Catalog No. 3384B6200
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3384B6200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 3384B6200 Supplier R&D Systems™ Supplier No. 3384B6200
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

Measured by its binding ability in a functional ELISA. Immobilized rhEphB6/Fc Chimera at 2 µg/mL (100 µL/well) can bind rmEphrin-B2/Fc Chimera with a linear range of 0.0780-5.00 ng/mL.

Specifications

Accession Number AAI10608.2
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 2051
Molecular Weight (g/mol) M.W. Observed: 104 kDa, under reducing conditions.
Name EphB6 Fc Chimera
Quantity 200 μg
Source Mouse myeloma cell line, NS0-derived human EphB6 protein Human EphB6 (Leu17-Ser579) (N-terminus) Accession #AAI10608.2 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.