Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human ERAF Protein
SDP

Catalog No. NBP130205 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP130205 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP130205 Supplier Novus Biologicals™ Supplier No. NBP130205
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-102 of Human ERAF The Recombinant Human ERAF Protein is derived from E. coli. The Recombinant Human ERAF Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_057717
For Use With (Application) ELISA, Western Blot
Formulation 20mM Tris-HCl buffer (pH8.0), 10% glycerol
Gene ID (Entrez) 51327
Molecular Weight (g/mol) 11.8kDa
Name ERAF Protein
Purification Method Protein
Quantity 0.1 mg
Immunogen MALLKANKDL ISAGLKEFSV LLNQQVFNDP LVSEEDMVTV VEDWMNFYIN YYRQQVTGEP QERDKALQEL RQELNTLANP FLAKYRDFLK SHELPSHPPP SS
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.