Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human ErbB2/Her2 Fc Chimera Protein
SDP

Catalog No. 1129ER01M Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-29E-R01M 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-29E-R01M Supplier R&D Systems™ Supplier No. 1129ER01M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human ErbB2/Her2 Fc Chimera Protein is derived from NS0. The Recombinant Human ErbB2/Her2 Fc Chimera Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number NP_004439
Characterization Bioactivity
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 2064
Molecular Weight (g/mol) 96.8kDa (monomer)
Quantity 1 mg
Source Mouse myeloma cell line,NS0-derived human ErbB2/Her2 protein, (N-terminus) Human Erb B2 (Thr23-Thr652) Accession &Num; NP_004439 IEGRMD Human IgG1 (Pro100-Lys330) 6 His-tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10EU per 1μg of the protein by the LAL method.
Gene Symbol ERBB2
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 100μg/mL in sterile PBS.
Purity or Quality Grade >90%, by SDS-PAGE with silver staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.