Learn More
R&D Systems™ Recombinant Human ErbB3/Her3 Fc Chimera Avi-tag Protein
Description
Specifications
Specifications
| Accession Number | P21860.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 2065 |
| Molecular Weight (g/mol) | M.W.-observed: 110-130 kDa, under reducing conditions, M.W.-theroretical: 97 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human ErbB3/Her3 protein Human ErbB3/Her3 (Ser20-Thr643) (c-terminus) Accession # P21860.1 IEGRMD Human IgG1 (Pro100-Lys330) Avi-tag N-terminus C-terminμs |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.