Learn More
Description
Specifications
Specifications
| For Use With (Application) | SDS-PAGE |
| Formulation | Phosphate buffered saline (pH7.4) containing 10% glycerol, 1mM DTT |
| Gene ID (Entrez) | 55224 |
| Molecular Weight (g/mol) | TMW: 47.2kDa |
| Quantity | 0.01 mg |
| Storage Requirements | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
| Gene Alias | EC 2.7.1.82, EKI 2, EKI2HMFT1716, ethanolamine kinase 2, Ethanolamine kinase-like protein, FLJ10761 |
| Common Name | ETNK2 |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
For research use only and is not approved for use in humans or in clinical diagnosis
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
