Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human FABP3/H-FABP Protein

Catalog No. 9479CL100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
94-79C-L100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 94-79C-L100 Supplier R&D Systems™ Supplier No. 9479CL100
Add to Cart
Add to Cart
This item is not returnable. View return policy

R&D Systems™ Recombinant Human FABP3/H-FABP Protein CF is a member of a large superfamily of lipid binding proteins that are expressed in a tissue specific manner.

The Recombinant Human FABP3/H-FABP Protein is derived from E. coli.

Specifications

Accession Number P05413
Characterization Bioactivity not tested
For Use With (Application) Flow Cytometry
Formulation Lyophilized from a 0.2μm filtered solution in PBS, EDTA and DTT.
Gene ID (Entrez) 2170
Molecular Weight (g/mol) 16kDa
Quantity 100 μg
Source E. coli-derived human FABP3/H-FABP protein, (N-terminus) MHHHHHH GS Human FABP3/H-FABP (Met1-Ala133) Accession &Num; P05413 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10EU per 1μg of the protein by the LAL method.
Gene Alias FABP11, fatty acid binding protein 11, fatty acid binding protein 3, muscle and heart (mammary-derived growthinhibitor), Fatty acid-binding protein 3, Fatty acid-binding protein 3, muscle, fatty acid-binding protein, heart, Heart-type fatty acid-binding protein, HFABP, H-FABP, H-FABPM-FABP, Mammary-derived growth inhibitor, MDGI, Muscle fatty acid-binding protein, O-FABP
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 1mg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.