Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human FAM20B Protein

Catalog No. 8427FM050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
84-27FM050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 84-27FM050 Supplier R&D Systems™ Supplier No. 8427FM050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to enhance survival of ATDC5 mouse chondrogenic cells. The ED50 for this effect is 0.8-4 μg/mL. Measured by its ability to phosphorylate D-xylose. The specific activity is >15 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number O75063
For Use With (Application) Bioactivity, Enzyme Activity
Formulation Lyophilized from a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 9917
Molecular Weight (g/mol) M.W. (Observed): 44-53 kDa, reducing conditions; M.W. (theoretical): 44 kDa
Name FAM20B
Preparation Method Reconstitute at 500μg/mL in 25mM Tris-HCl, pH7.5.
Quality Control Testing SDS-PAGE: 44-53kDa, reducing conditions
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human FAM20B protein Asn27-Leu409, with an N-terminal HA tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Tyr
Without Additives Carrier Protein
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol FAM20B
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL in 25 mM Tris-HCl, pH7.5.
Purity or Quality Grade >95%, by SDS-PAGE with silver staining
Protein FAM20B
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.