Learn More
R&D Systems™ Recombinant Human FAM20C Protein
Description
Order Info
The product is shipped at ambient temperature.
Specifications
Specifications
| Accession Number | Q8IXL6 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 56975 |
| Molecular Weight (g/mol) | M.W. (Observed): 70-82 kDa, reducing conditions; M.W. (theoretical): 60 kDa |
| Name | dentin matrix protein 4, DKFZp547C074, DMP4, DMP-4, family with sequence similarity 20, member C, GEF-CK, RNS |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived human FAM20C protein Val63-Arg584 with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Regulatory Status | RUO |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.