Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Fas/TNFRSF6/CD95 Fc Chimera Protein
SDP

Catalog No. 326FS050 Shop All R&D Systems Products
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
326FS050 Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
326FS050CF Lyophilized from a 0.2μm filtered solution in PBS.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 326FS050 Supplier R&D Systems™ Supplier No. 326FS050
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

New rhFas! HEK293 expressed; ∽2 fold better activity; Lower endotoxin specification; No His tag!

The Recombinant Human Fas/TNFRSF6/CD95 Fc Chimera Protein is derived from NS0. The Recombinant Human Fas/TNFRSF6/CD95 Fc Chimera Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 355
Molecular Weight (g/mol) 45 kDa (monomer)
Name Fas/TNFRSF6/CD95 Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived human Fas/TNFRSF6/CD95 protein, (N-terminus) Human Fas (Arg17-Asn173) Accession &Num; Q5T9P3 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.