Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Fc gamma RIIIA/CD16a (V176F) Protein

Catalog No. 8894FC050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8894FC050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8894FC050 Supplier R&D Systems™ Supplier No. 8894FC050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Fc gamma RIIIA/CD16a (V176F) Protein is derived from HEK293. The Recombinant Human Fc gamma RIIIA/CD16a (V176F) Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P08637
For Use With (Application) Bioactivity
Gene ID (Entrez) 2214
Molecular Weight (g/mol) Observed M.W.: 43-58kDa, reducing conditions, Theoretical M.W.: 23kDa
Quantity 50 μg
Source Human embryonic kidney cell,HEK293-derived human Fc gamma RIIIA/CD16a protein Gly17-Gln208,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias CD16a, CD16a antigen, CD16FCRIIIA, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), Fc fragment of IgG, low affinity IIIa, receptor for (CD16), Fc gamma receptor III-A, FCG3, Fc-gamma receptor III-2 (CD 16), Fc-gamma receptor IIIb (CD16), Fc-gamma RIII, Fc-gamma RIIIa, Fc-gamma RIII-alpha, FCGR3, FCGR3A, FCGRIII, FcgRIIIA, FCR-10, FcRIII, FcRIIIa, IGFR3FcgammaRIIIA, immunoglobulin G Fc receptor III, low affinity III, receptor for (CD16), low affinity immunoglobulin gamma Fc region receptor III-A, neutrophil-specific antigen NA
Common Name Fc gamma RIIIA/CD16a
Gene Symbol FCGR3A
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Human Fc gamma RIIIa/CD16a (V176F) was immobilize on a Ms x PolyHis coated plate, it binds biotinylated Human IgG. The concentration of biotinylated Human IgG that produces 50% of the optimal binding response is approximately 0.5-2.5μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Gly17-Gln208, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.