Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Fc gamma RIIIA (F176) His Avi Protein

Catalog No. AVI8894050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AVI8894050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AVI8894050 Supplier R&D Systems™ Supplier No. AVI8894050
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Recombinant Human Fc gamma RIIIA (F176) His Avi Protein is derived from HEK293

  • Recombinant Protein
  • Carrier Free (CF)
  • The carrier free version does not contain Bovine Serum Albumin (BSA)

Specifications

Accession Number P08637-1
For Use With (Application) Bioactivity
Gene ID (Entrez) 2214
Molecular Weight (g/mol) 25 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 50 μg
Source

Human embryonic kidney cell, HEK293-derived human Fc gamma RIIIA/CD16a protein

Human Fc gamma RIIIA (F176) (Gly17-Gln208) Accession # P08637-1HHHHHHHAvi-tagN-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Biotin
Reconstitution Reconstitute at 200 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose
Activity The biotin to protein ratio is greater than 0.7 as determined by the HABA assay. Measured by its binding ability in a functional ELISA. When Human IgG is immobilized at 5 μg/mL (100 μL/well), Biotinylated Recombinant Human Fc gamma RIIIA/CD16a (F176) His-tag Avi-tag (Catalog # AVI8894) binds with an ED50 of 50-300 ng/mL.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.