Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Fc gamma RIIIB/CD16b (aa18-200) Protein
SDP

Catalog No. 9948FC100 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
99-48FC-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 99-48FC-100 Supplier R&D Systems™ Supplier No. 9948FC100

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Fc gamma RIIIB/CD16b (aa18-200) Protein is derived from HEK293. The Recombinant Human Fc gamma RIIIB/CD16b (aa18-200) Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number NP_000561
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 2215
Molecular Weight (g/mol) 22 kDa
Quantity 100 μg
Source Human embryonic kidney cell, HEK293-derived human Fc gamma RIIIB/CD16b protein Met18-Ser200, with a C-Terminal 6-His tag
Storage Requirements 12 months from date of receipt, ≤ -20°C as supplied; 1 month, 2 to 8°C under sterile conditions after reconstitution; 3 months, ≤ -20°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Cross Reactivity Fc gamma RIIIB/CD16b
Species Human
Reconstitution Reconstitute at 500 μg/mL in PBS.
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.