Learn More
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT with BSA as a carrier protein. |
| Gene ID (Entrez) | 2255 |
| Molecular Weight (g/mol) | Observed Molecular Wt.: 19-22 kDa, reducing conditions |
| Name | FGF-10 |
| Quantity | 25 μg |
| Source | E. coli-derived human FGF-10 protein Cys37-Ser208 & Gly41-Ser208 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | FGF10, FGF-10, fibroblast growth factor 10, Keratinocyte growth factor 2, KGF2, KGF-2, produced by fibroblasts of urinary bladder lamina propria |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
