Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human FGF-4 Protein

Catalog No. 235F401M
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-5F4-01M 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-5F4-01M Supplier R&D Systems™ Supplier No. 235F401M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human FGF-4 Protein is derived from E. coli. The Recombinant Human FGF-4 Protein has been validated for the following applications: Bioactivity.

Order Info

Ships at ambient temperature.

Specifications

Accession Number P08620
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein
Gene ID (Entrez) 2249
Molecular Weight (g/mol) 17 kDa
Quantity 1 mg
Source E. coli-derived human FGF-4 protein Ser54-Leu206
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.01 EU per 1μg of the protein by the LAL method
Gene Alias FGF4, fibroblast growth factor 4, HBGF-4
Gene Symbol FGF4
Biological Activity 0.25 to 1.25ng/mL
Product Type Recombinant protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.