Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human FGF basic Heat Stable Protein, Animal-Free
SDP

Catalog No. RD10000340 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10000340 25 μg
RD10000339 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. RD10000340 Supplier R&D Systems™ Supplier No. BTFGFBHSAFL025

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells. Raines, E.W. et al. (1985) Methods Enzymol. 109:749. The ED50 for this effect is 0.0500-0.600 ng/mL. The specific activity of Animal-Free™ Recombinant Human FGF basic Heat Stable is >1.60 x 106 IU/mg, which is calibrated against the human FGF basic/FGF2 WHO International Standard (NIBSC code: 90/712).

Specifications

Accession Number NP_001997.5
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in HEPES and Sodium Sulfate with Trehalose.
Gene ID (Entrez) 2247
Molecular Weight (g/mol) MolecularWeight-observed: 16-19 kDa, under reducing conditions, MolecularWeight-theroretical: 17 kDa
Quantity 25 μg
Source E. coli-derived human FGF basic/FGF2/bFGF protein Ala135-Ser288 (with modifications) Produced using non-animal reagents in an animal-free laboratory.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin
Conjugate Unconjugated
Reconstitution Reconstitute the 25 μg size at 250 μg/mL in sterile water. Reconstitute all other sizes at 500 μg/mL in sterile water.
Product Line R&D Systems
Protein FGF basic/FGF2/bFGF
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.