Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human FGFR1 Kinase Domain Protein
SDP

Catalog No. 8657FR020 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
86-57F-R020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 86-57F-R020 Supplier R&D Systems™ Supplier No. 8657FR020
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human FGF R1 Kinase Domain Protein is derived from Sf 21 (baculovirus). The Recombinant Human FGF R1 Kinase Domain Protein has been validated for the following applications: Enzyme Activity.

Specifications

Accession Number P11362
For Use With (Application) Enzyme Activity
Gene ID (Entrez) 2260
Molecular Weight (g/mol) Observed M.W.: 45-58kDa, reducing conditions, Theoretical M.W.: 49kDa
Name FGFR1
Quantity 20 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived human FGF R1 protein Met399-Arg822 with an N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method.
Gene Alias basic fibroblast growth factor receptor 1, BFGFR, bFGF-R-1, CD331, CD331 antigen, CEK, EC 2.7.10, EC 2.7.10.1, FGF R1, FGFBR, FGFR-1, fibroblast growth factor receptor 1, FLGH3, FLJ99988, Flt-2, FLT2H4, Fms-like tyrosine kinase 2, fms-related tyrosine kinase 2, H2, H5, HBGFR, heparin-binding growth factor receptor, hydroxyaryl-protein kinase, KAL 2, KAL2, N-SAM, OGD, Proto-oncogene c-Fgr, soluble FGFR1 variant 1, soluble FGFR1 variant 2
Common Name FGF R1
Gene Symbol FGFR1
Biological Activity Measured by its ability to hydrolyze the 5’-phosphate groups from the substrate adenosine-5’-triphosphate (ATP). The orthophosphate product is measured by a Malachite Green Phosphate Detection Kit. The specific activity is >12 pmol/min/μg, as measured under the described conditions. See Activity Assay Protocol on RnDSystems.com.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Spodoptera frugiperda, Sf 21 (baculovirus)-derived Met399-Arg822 with an N-terminal Met and 6-His tag
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5μg per lane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.