Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Fibrillin-1/FBN1 Fc Chimera Protein

Catalog No. 10224FI050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-224-FI050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-224-FI050 Supplier R&D Systems™ Supplier No. 10224FI050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measured by its binding ability in a functional ELISA.

When Recombinant Human MFAP4 is immobilized at 0.5 μg/mL (100 μL/well), the concentration of Recombinant Human Fibrillin-1/FBN1 Fc Chimera that produces 50% of the optimal binding response is 0.1 to 0.6 μg/mL.

Specifications

Accession Number P35555
For Use With (Application) Bioactivity
Gene ID (Entrez) 2200
Molecular Weight (g/mol) 95 kDa
Name ACMICD, Asprosin, FBN, Fibrillin 1, fibrillin 1 (Marfan syndrome), fibrillin 15, fibrillin-1, GPHYSD2, MASS, MFS1, OCTD, SGS, SSKS, WMS, WMS2
Quantity 50 μg
Source

Mouse myeloma cell line, NS0-derived human Fibrillin-1/FBN1 protein

Human Fibrillin-1 (Ala25-Thr660) Accession # P35555IEGRMDHuman IgG1 (Pro100-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 500 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.