Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human FLAG-WWP1 Protein, CF

Catalog No. E3232050
Change view
Click to view available options
50 ug
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
E3232050 50 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E3232050 Supplier R&D Systems™ Supplier No. E3232050
This item is not returnable. View return policy

Recombinant Human WWP1 is a Ubiquitin Ligase (E3) that functions downstream of a Ubiquitin-activating (E1) enzyme and a Ubiquitin-conjugating (E2) enzyme to conjugate Ubiquitin to substrate proteins. Reaction conditions will need to be optimized for each specific application.

Specifications

Accession Number Q9H0M0
For Use With (Application) Bioactivity
Gene ID (Entrez) 11059
Molecular Weight (g/mol) 107 kDa
Name AIP5, AIP5Nedd-4-like ubiquitin-protein ligase, atrophin-1 interacting protein 5, Atrophin-1-interacting protein 5, DKFZP434D2111, EC 6.3.2, EC 6.3.2.-, hSDRP1, NEDD4-like E3 ubiquitin-protein ligase WWP1, TGIF-interacting ubiquitin ligase 1, TIUL1, Tiul11, WW domain containing E3 ubiquitin protein ligase 1, WW domain-containing protein 1
Quantity 50 μg
Source Trichoplusia ni, T. ni (baculovirus)-derived human WWP1 protein 2X FLAG TM tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 48 months from date of receipt, -70 °C as supplied. 3 months, -70 °C under sterile conditions after opening.
Conjugate Unconjugated
Species Human
Buffer X mg/ml (X μM) in 50 mM HEPES pH 7.5, 200 mM NaCl, 10% (v/v) Glycerol, 1 mM DTT
Purity or Quality Grade >85%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.