Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Flt-3 Ligand, ACFP Protein

Catalog No. ACFP308
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ACFP308 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. ACFP308 Supplier R&D Systems™ Supplier No. ACFP308
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Flt-3 Ligand, ACFP Protein is derived from Sf 9 (baculovirus). The Recombinant Human Flt-3 Ligand, ACFP Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number AAA17999.1
For Use With (Application) Bioactivity
Gene ID (Entrez) 2323
Molecular Weight (g/mol) 17.5 kDa
Quantity 50 μg
Source Spodoptera frugiperda,Sf 9 (baculovirus)-derived human Flt-3 Ligand/FLT3L protein Thr27-Pro184 Produced in an animal component free process (ACFP).
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.01 EU per 1 μg of the protein by the LAL method.
Gene Alias FL, FLG3L, Flt3 ligand, Flt3L, FLT3LG, fms-related tyrosine kinase 3 ligand, SL cytokine
Gene Symbol FLT3LG
Specific Reactivity Flt-3 Ligand
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Purity or Quality Grade >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.