Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Flt-3 Ligand/FLT3L GMP Protein, CF
SDP

Catalog No. RD10008962 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10008962 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10008962 Supplier R&D Systems™ Supplier No. BTFT3LGMP050
Add to Cart
Add to Cart

Recombinant Protein

Measured in a cell proliferation assay using OCI-AML5 acute myeloid leukemia cells. The ED50 for this effect is 0.200-2.00 ng/mL. The specific activity of Recombinant Human Flt-3 Ligand/FLT3L is >5.00 x 105 units/mg, which is calibrated against the human Flt-3 Ligand WHO standard (NIBSC code: 96/532).

Specifications

Accession Number P49771.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 2323
Molecular Weight (g/mol) Observed: 17 kDa, under reducing conditions., Observed: 18 kDa
Quantity 50 μg
Source E. coli-derived human Flt-3 Ligand/FLT3L protein Thr27 - Ala181 Produced using non-animal reagents in an animal-free laboratory. Manufactured and tested under cGMP guidelines.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. A minimum of 12 months when stored at ≤ -20°C as supplied. Refer to lot specific COA for the Use by Date. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, ≤ -20°C under sterile conditions after reconstitution.
Regulatory Status GMP
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias FL, FLG3L, Flt3 ligand, Flt3L, FLT3LG, fms-related tyrosine kinase 3 ligand, SL cytokine
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >97%, by SDS-PAGE with quantitative densitometry by Coomassie™ Blue Staining. The molecular weight by mass spectrometry is 17731 Da ± 50 Da, with a second mass of 17600 Da ± 50 Da also present
Protein Flt-3 Ligand/FLT3L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.