Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Flt-3 Ligand/FLT3L (HEK293) Protein

Catalog No. 308FKHB050
Change view
Click to view available options
Quantity:
1 mg
10 μg
250 μg
50 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
308FKHB050 50 μg
308FKHB010 10 μg
308FKHB01M 1 mg
308FKHB250 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 308FKHB050 Supplier R&D Systems™ Supplier No. 308FKHB050
Add to Cart
Add to Cart
This item is not returnable. View return policy
Measured in a cell proliferation assay using BaF3 mouse pro-B cells transfected with mouse Flt-3. The ED50 for this effect is 0.150-1.50 ng/mL.

Specifications

Accession Number AAA17999.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 2323
Molecular Weight (g/mol) M.W.-observed: 22-31 kDa, under reducing conditions., M.W.-theroretical: 18 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human Flt-3 Ligand/FLT3L protein Thr27-Pro185
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias FL, FLG3L, Flt3 ligand, Flt3L, FLT3LG, fms-related tyrosine kinase 3 ligand, SL cytokine
Conjugate Unconjugated
Reconstitution Reconstitute the 10 μg size at 100 μg/mL in PBS. Reconstitute all other sizes (at 250 μg/mL in PBS.)
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Flt-3 Ligand/FLT3L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.