Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Follistatin-like 5/FSTL5 Protein

Catalog No. 9066FN100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-66F-N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 90-66F-N100 Supplier R&D Systems™ Supplier No. 9066FN100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Follistatin-like 5/FSTL5 Protein is derived from CHO. The Recombinant Human Follistatin-like 5/FSTL5 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number AAH36502
For Use With (Application) Bioactivity
Gene ID (Entrez) 56884
Molecular Weight (g/mol) Observed M.W.: 86-102kDa, reducing conditions, Theoretical M.W.: 94kDa
Quantity 100 μg
Source Chinese Hamster Ovary cell line,CHO-derived human Follistatin-like 5/FSTL5 protein Arg21-Ala847,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias Follistatin like 5, follistatin-like 5, Follistatin-like protein 5, follistatin-related protein 5, FSL5, FSTL5, KIAA1263DKFZp566D234
Common Name FSTL5
Gene Symbol FSTL5
Biological Activity Measured by its ability to inhibit Hep3B human hepatocellular carcinoma cell survival. The ED50 for this effect is typically 3-12μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Arg21-Ala847, with a C-terminal 6-His tag
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.