Learn More
R&D Systems™ Recombinant Human FSH alpha/beta Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 2488 |
| Molecular Weight (g/mol) | 10.2 kDa (FSH alpha) & 12.4 kDa (FSH beta) |
| Name | FSH alpha/beta |
| Quantity | 10 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human FSH alpha/beta protein, (N-terminus) Human FSH alpha Ala25-Ser116 Accession &Num; P01215 Human FSH beta Cys21-Glu129 Accession &Num; P01225 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Noncovalently-linked heterodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.