Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Fucosyltransferase 9/FUT9 Protein
SDP

Catalog No. 9347GT020 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
93-47G-T020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 93-47G-T020 Supplier R&D Systems™ Supplier No. 9347GT020

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

For Research Use Only

FUT9 has been formulated so that it can be used in the cell surface glycoengineering of living cells, and does not affect cell viability or native phenotype apart from the intended impact on cell glycobiology.

Measured by its ability to transfer fucose from GDP-fucose to N -Acetyllactosamine The specific activity is >1,000 pmol/min/μg, as measured under the described conditions.

Order Info

Ships at ambient temperature.

Specifications

Accession Number Q9Y231
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 10690
Molecular Weight (g/mol) M.W. (Observed): 40-50 kDa, reducing conditions; M.W. (theoretical): 39 kDa
Quantity 20 μg
Source Chinese Hamster Ovary cell line, CHO-derived human Fucosyltransferase 9/FUT9 protein Thr33-Asn359
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Product Type Recombinant protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein FUT9
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.