Learn More
Description
Specifications
Specifications
| Accession Number | P35637 |
| For Use With (Application) | SDS-PAGE, Western Blot |
| Formulation | 25mM Tris pH 8.0, 250mM NaCl, 8 M Urea, 1mM DTT |
| Gene ID (Entrez) | 2521 |
| Name | FUS Protein |
| Purification Method | This FUS Protein has a Purity >95%. This product was tested in Western Blot using NB100-562. The antibody was used to detect 60ng on the western blot. |
| Quantity | 10 μg |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Product Line | Novus |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
