Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human G-CSF Protein

Catalog No. 214CS005
Change view
Click to view available options
Quantity:
25 μg
5 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetic Acid with BSA as a carrier protein.
Supplied as a 0.2μm filtered solution in Acetic Acid.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
214CS005 5 μg Lyophilized from a 0.2μm filtered solution in Acetic Acid with BSA as a carrier protein.
214CS025 25 μg Lyophilized from a 0.2μm filtered solution in Acetic Acid with BSA as a carrier protein.
214CS005CF 5 μg Supplied as a 0.2μm filtered solution in Acetic Acid.
214CS025CF 25 μg Supplied as a 0.2μm filtered solution in Acetic Acid.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 214CS005 Supplier R&D Systems™ Supplier No. 214CS005
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human G-CSF Protein is derived from E. coli. The Recombinant Human G-CSF Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Acetic Acid with BSA as a carrier protein.
Gene ID (Entrez) 1440
Molecular Weight (g/mol) 18.8 kDa
Name G-CSF
Quantity 5 μg
Source E. coli-derived human G-CSF protein Thr31-Pro204,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.