Learn More
R&D Systems™ Recombinant Human G-CSF Protein
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Acetic Acid. |
| Gene ID (Entrez) | 1440 |
| Molecular Weight (g/mol) | 18.8 kDa |
| Name | G-CSF |
| Quantity | 5 μg |
| Source | E. coli-derived human G-CSF protein Thr31-Pro204,with an N-terminal Met |
| Storage Requirements | Do not freeze. 6 months from date of receipt, 2 to 8° C as supplied. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.