Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human G protein alpha inhibitor 1 His Protein
SDP

Catalog No. NBP17246650 Shop All R&D Systems Products
Change view
Click to view available options
50μg; Unlabeled
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NBP17246650 50μg; Unlabeled
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP17246650 Supplier Novus Biologicals™ Supplier No. NBP17246650UG
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-354 of Human G protein alpha inhibitor 1 The Recombinant Human G protein alpha inhibitor 1 Protein is derived from E. coli. The Recombinant Human G protein alpha inhibitor 1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_002060
For Use With (Application) SDS-PAGE
Formulation Liquid. 20mM Tris-HCl buffer (pH8.0) containing 10% glycerol
Gene ID (Entrez) 2770
Molecular Weight (g/mol) 42.7kDa
Name G protein alpha inhibitor 1 Protein
Purification Method SDS-PAGE
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.