Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human G6PD His-tag Protein

Catalog No. 10096DH050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10096DH050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10096DH050 Supplier R&D Systems™ Supplier No. 10096DH050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to dehydrogenate glucose-6-phosphate. The specific activity is >14,000 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number P11413
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NADP and Glycerol.
Gene ID (Entrez) 2539
Molecular Weight (g/mol) M.W. (Observed): 56 kDa, reducing conditions; M.W. (theoretical): 60 kDa
Quantity 50 μg
Source E. coli-derived human G6PD protein Ala2-Leu515, with C-terminal 6 x His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Specific Reactivity Glucose 6 Phosphate Dehydrogenase
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Buffer Supplied as a 0.2 μm filtered solution in Tris, NADP and Glycerol.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Protein Glucose 6 Phosphate Dehydrogenase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.