Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Galectin-3BP/MAC-2BP Protein

Catalog No. 2226GAB050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-26G-AB050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-26G-AB050 Supplier R&D Systems™ Supplier No. 2226GAB050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Galectin-3BP/MAC-2BP Protein is derived from NS0. The Recombinant Human Galectin-3BP/MAC-2BP Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q08380.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS and DTT with Trehalose.
Gene ID (Entrez) 3959
Molecular Weight (g/mol) 65 kDa
Name 90K, Basement membrane autoantigen p105, BTBD17B, galectin-3-binding protein, Galectin3BP, L3 antigen, Lectin galactoside-binding soluble 3-binding protein, lectin, galactoside-binding, soluble, 3 binding protein, LGALS3BP, M2BP, Mac-2 BP, Mac-2-binding protein, MAC2BP, MAC-2BP, MAC-2-BP, serum protein 90K, Tumor-associated antigen 90K
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human Galectin-3 BP/MAC-2 BP protein Val19-Asp585, with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 500μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.