Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Galectin-9 Protein
SDP

Catalog No. 9064GA050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-64G-A050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 90-64G-A050 Supplier R&D Systems™ Supplier No. 9064GA050
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

E.coli expressed rh Galectin-9 (Catalog # 2045-GA) is also available with high TIM-3 binding affinity!

Measured by its ability of the immobilized protein to support the adhesion of Jurkat human acute T cell leukemia cells. The ED50 for this effect is 0.75-4.5 μg/mL.

Specifications

Accession Number NP_002299
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl, EDTA, DTT and Trehalose.
Gene ID (Entrez) 3965
Molecular Weight (g/mol) M.W. (Observed): 42-51 kDa, reducing conditions; M.W. (theoretical): 36 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human Galectin-9 protein Met1-Thr323
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias ecalectin, gal-9, GAL9, galectin 9, galectin-9, Galectin9, HUAT, lectin, galactoside-binding, soluble, 9, LGALS9, LGALS9A, MGC117375, MGC125973, MGC125974, Tumor antigen HOM-HD-21, urate transporter/channel protein
Common Name Galectin-9
Gene Symbol LGALS9
Biological Activity Measured by its ability of the immobilized protein to support the adhesion of Jurkat human acute T cell leukemia cells. The ED50 for this effect is typically 0.75-4.5μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 200 μg/mL in PBS.
Expression System Human embryonic kidney cell, HEK293-derived Met1-Thr323
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Galectin-9
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.